missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC13A3/NaDC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69630
This item is not returnable.
View return policy
Description
SLC13A3/NaDC3 Polyclonal specifically detects SLC13A3/NaDC3 in Human samples. It is validated for Western Blot.
Specifications
| SLC13A3/NaDC3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| hNaDC3, Na(+)/dicarboxylate cotransporter 3, NaDC-3, NADC3SDCT2naDC-3, sodium-dependent high affinity dicarboxylate transporter 3, Sodium-dependent high-affinity dicarboxylate transporter 2, solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3, solute carrier family 13 member 3 | |
| Rabbit | |
| 67 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8WWT9 | |
| SLC13A3 | |
| Synthetic peptides corresponding to SLC13A3(solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3) The peptide sequence was selected from the N terminal of SLC13A3 (NP_073740). Peptide sequence MAVYWCTEALPLSVTALLPIVLFPFMGILPSNKVCPQYFLDTNFLFLSGL The peptide sequence for this immunogen was taken from within the described region. | |
| Protein A purified | |
| RUO | |
| 64849 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction