missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC13A1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA564056

Product Code. 15761822

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: QIINAEAEVE ATQMTYFNGS TNHGLEIDES VNGHEINERK EKTKPVPGYN N Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 67%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC13A1
Polyclonal
Unconjugated
Slc13a1
AI314066; AI987826; Na(+)/sulfate cotransporter; NAS1; NASI1; NaSi-1; Renal sodium/sulfate cotransporter; SLC13A1; solute carrier family 13 (sodium/sulfate symporters), member 1; solute carrier family 13 (sodium/sulphate symporters), member 1; solute carrier family 13 member 1
Rabbit
Antigen affinity chromatography
RUO
6561
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BZW2
Slc13a1
Recombinant protein corresponding to Human SLC13A1. Recombinant protein control fragment (Product #RP-103465).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.