missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC12A4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15927-1-AP-20UL
This item is not returnable.
View return policy
Description
SLC12A4 mediates electroneutral potassium-chloride cotransport when activated by cell swelling. SLC12A4 may contribute to cell volume homeostasis in single cells.Specifications
| SLC12A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC12A4 | |
| Erythroid K Cl cotransporter 1, hKCC1, KCC1, SLC12A4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20498, 29501, 6560 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q63632, Q9JIS8, Q9UP95 | |
| SLC12A4 | |
| SLC12A4 Fusion Protein Ag8762 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction