missing translation for 'onlineSavingsMsg'
Learn More

SLC12A3 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18421223 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421223 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421223 Supplier Novus Biologicals Supplier No. NBP144270G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC12A3 Polyclonal antibody specifically detects SLC12A3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC12A3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FLJ96318, Na-Cl symporter, NCCT, solute carrier family 12 (sodium/chloride transporters), member 3, solute carrier family 12 member 3, thiazide-sensitive Na-Cl cotransporter, Thiazide-sensitive sodium-chloride cotransporter, TSCNaCl electroneutral thiazide-sensitive cotransporter
Host Species Rabbit
Immunogen Produced against a synthetic peptide mapping to a segment of rat NCC (amino acids 74-95), N-terminal
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6559
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.