missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC11A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112711
This item is not returnable.
View return policy
Description
SLC11A1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
NRAMP1, also known as SLC11A1, is a member of the solute carrier family 11 (proton-coupled divalent metal ion transporters) family and encodes a multi-pass membrane protein. The protein functions as a divalent transition metal (iron and manganese) transporter involved in iron metabolism and host resistance to certain pathogens. Mutations in this gene have been associated with susceptibility to infectious diseases such as leprosy and tuberculosis, and inflammatory diseases such as Crohn disease and rheumatoid arthritis. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
Specifications
| SLC11A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC11A1 | |
| Bcg; host resistance locus Bcg/Ity/Lsh; integral membrane protein; Itg; Ity; Lsh; natural resistance associated macrophage protein; natural resistance associated macrophage protein 1; natural resistance-associated macrophage protein; natural resistance-associated macrophage protein 1; natural resistance-associated macrophages protein 1; NRAMP; NRAMP 1; Nramp1; Slc11a1; solute carrier 11 family member 1; solute carrier family 11 (proton-coupled divalent metal ion transporter), member 1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1; solute carrier family 11 (sodium/phosphate symporters), member 1; solute carrier family 11 A1; solute carrier family 11 member 1; solute carrier family 11 member a1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 316519, 6556 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P49279, P70553 | |
| SLC11A1 | |
| Recombinant Human Natural resistance-associated macrophage protein 1 protein (1-58AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction