missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC10A2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-36987
This item is not returnable.
View return policy
Description
SLC10A2 Polyclonal specifically detects SLC10A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| SLC10A2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Apical sodium-dependent bile acid transporter, ASBT, IBAT, ileal apical sodium-dependent bile acid transporter, Ileal Na(+)/bile acid cotransporter, ileal sodium/bile acid cotransporter, Ileal sodium-dependent bile acid transporter, ISBT, Na(+)-dependent ileal bile acid transporter, NTCP2PBAM, Sodium/taurocholate cotransporting polypeptide, ileal, solute carrier family 10 (sodium/bile acid cotransporter family), member 2, Solute carrier family 10 member 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6555 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 3 - 6 ug/ml, Immunohistochemistry-Paraffin 3 - 6 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_000443.1 | |
| SLC10A2 | |
| Peptide with sequence C-YKANGGFQPDEK corresponding to C-Terminus according to NP_000443.1. | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction