missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC10A1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93184-0.02ml
This item is not returnable.
View return policy
Description
SLC10A1 Polyclonal antibody specifically detects SLC10A1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| SLC10A1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Cell growth-inhibiting gene 29 protein, Na(+)/bile acid cotransporter, Na(+)/taurocholate transport protein, Na/taurocholate cotransporting polypeptide, NTCP, NTCP1, NTCPgrowth-inhibiting protein 29, sodium/bile acid cotransporter, sodium/taurocholate cotransporter, Sodium/taurocholate cotransporting polypeptide, solute carrier family 10 (sodium/bile acid cotransporter family), member 1, Solute carrier family 10 member 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human SLC10A1 (NP_003040.1). VAVTVLSAINVGKSIMFAMTPLLIATSSLMPFIGFLLGYVLSALFCLNGRCRRTVSMETGCQNVQLCSTILNVAFPPEVIGPLFFFPLLYMIFQLGEGLLLIAIFWCYEKFKTPKDKTKMIYTAATTEETIPGALGNGTYKGEDCSPCTA | |
| 0.02 mL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6554 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction