Learn More
Invitrogen™ SLA/LP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596578
Description
Immunogen sequence: STTSCFAPRV PDRLEELAVI CANYDIPHIV NNAYGVQSSK CMHLIQQGAR VGRIDAFVQS LDKNFMVPVG GAIIAGFNDS FIQEISKMYP GRASASPSLD VLITLLSLGS NGYKKLLKER KEMFSYLSNQ IKKLSEAYNE RLLHTPHNPI SLAMTLKTLD EHRDKAVTQL GSMLFTRQVS GARVVPLGSM QTVSGYTFRG FMSHTNNYPC AYLNAASAIG MKMQDVDLFI KRLDRCLKAV RKERSKESDD NYDKTEDVDI EEMALKLDNV LLDTYQDASS; Positive Samples: 293T, THP-1, Mouse kidney, Rat liver; Cellular Location: Cytoplasm.
SLA/LP (SEPSECS) converts O-phosphoseryl-tRNA(Sec) to selenocysteinyl-tRNA(Sec) required for selenoprotein biosynthesis.
Specifications
| SLA/LP | |
| Polyclonal | |
| Unconjugated | |
| SEPSECS | |
| 9130208G10; AA986712; D5Ertd135e; hypothetical protein LOC393123; liver-pancreas antigen; LP; O-phosphoseryl-tRNA(Sec) selenium transferase; PCH2D; Sec synthase; SecS; Selenocysteine synthase; selenocysteinyl-tRNA(Sec) synthase; Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase; SEPSECS; sep-tRNA:Sec-tRNA synthase; SLA; SLA/LP; SLA/LP autoantigen; sla/lpl; SLA-p35; Soluble liver antigen; soluble liver antigen/liver pancreas antigen; soluble liver antigen/liver pancreas antigen, like; Soluble liver antigen/liver pancreas antigen-like; tRNA(Ser/Sec)-associated antigenic protein; TRNP48; UGA suppressor tRNA-associated protein; UGA suppressor tRNA-associated protein homolog; zgc:55980 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 211006, 51091, 679383 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.57 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P6M7, Q9HD40 | |
| SEPSECS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 222-501 of human SEPSECS (NP_0586513). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.