missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLAM (SLAMF1) Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHSLAMF1

Code : NEW

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15415133

  • £544.00 / 1 set
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

The Human SLAM (SLAMF1) ELISA quantitates Hu SLAM in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu SLAM. Principle of the method The Human SLAM solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

CD150 (SLAM, Signaling Lymphocyte Activation Molecule) belongs to the immunoglobulin gene superfamily, and is involved in T cell stimulation. SLAM is constitutively expressed on peripheral blood memory T cells, T cell clones, immature thymocytes, and a proportion of B cells, and is rapidly induced on naive T cells after activation. Activated B cells express the membrane bound form, the soluble form and the cytoplasmic isoforms of SLAM. It is suggested that signalling through homophilic SLAM-SLAM binding during B to B and B to T cell interactions enhances the expansion and differentiation of activated B cells. Reports suggest that the extracellular domain of CD150 is the receptor for the measles virus and acts as a co-activator on T cells and B cells. Moreover, CD150 is thought to interact with SH2D1A and with PTPN11 via its cytoplasmic domain. Mutations CD150 gene may be associated with X-linked lympho-proliferative disease (XLP).
Specifications

Specifications

Q13291
60 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
6504
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Cell Cycle & Proliferation
2°C to 8°C
Human
4 hr. 45 min.
61.44-15,000 pg/mL
60pg/mL to 15,000pg/mL
Biotin
Other Proteins
This ELISA pair antibody detects human SLAM. Other species not determined.
Human
Colorimetric Microplate Reader
CD150,CDw150,SLAM
<10%
HRP
RUO
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
RP11-528G1.4, CD150, CDw150, SLAM
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers