Learn More
Bollé Safety Slam Safety Glasses
Product Code. 12679200
Change view
Click to view available options
Lens Tint:
Shade 1.7
Shade 3
Shade 5
Yellow
Lens Coating:
Anti-fog, Impact-resistant, Scratch-resistant
Impact-resistant, Scratch-resistant
Unit Size:
Each
Product Code. Lens Tint Lens Coating unitSize
12679200 Yellow Anti-fog, Impact-resistant, Scratch-resistant Each
12689200 Shade 1.7 Impact-resistant, Scratch-resistant Each
12699200 Shade 3 Impact-resistant, Scratch-resistant Each
12609210 Shade 5 Impact-resistant, Scratch-resistant Each
4 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12679200
Brand: Bollé Safety SLAPSJ
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Stylish, easy-to-wear, provides protection at all times, and offers unbeatable comfort.
  • Stylish.
  • Easy to wear.
  • Protection at all times.
  • Unbeatable comfort.
  • EN 166: Guaranteed minimum resistance of the protective eyewear against everyday risks including dropping the protective eyewear onto the ground, ageing by light exposure, exposure to heat or corrosion etc.
  • EN 170, Risks relating to radiation - UV protection: The protection filters ultraviolet radiation. This type of protection is particularly recommended during activities where the source of ultraviolet radiation is predominant, such as mercury vapor lamps, actinic or germicides. Levels of application: 2-1.2, 2C-1.2 Lens color: Clear, blue, yellow or green. Adding the code 'C' indicates good color recognition by the holder.
  • Mechanical risk F - Mechanical risk - Low energy impact:& Low energy impact, resists a 6 mm, 0.86 g ball at 45 m/s.
  • Mechanical risk T - High speed particles at extreme temperatures: The letter T, immediately after the mechanical strength symbol, authorizes use for high-speed particles at extreme temperatures.
  • Optical class: Class 1, continuous work, worn permanently.
  • CE, European standard: Frame marked to indicate meeting CE standard.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Impact-resistant, Scratch-resistant
Description Model SLAPSJ
Certifications/Compliance (Europe) EN166, EN170/2C-1.2, Mechanical risk F/T, Optical class 1, CE
Lens Tint Yellow
Product Type Safety glasses
For Use With (Application) Indoor/cloudy sky, protection, continuous wear
No. per Case 200/Cs.
No. per Pack 1/bag, 10/Pk.
Weight (Metric) 24 g
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection EN170/2C-1.2