missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAM/CD150 mouse anti-Human, Clone: 5B4G1, Proteintech
mouse Monoclonal Antibody
Brand: Proteintech 60043-1-IG-150UL
This item is not returnable.
View return policy
Description
CD150, designated SLAM (Signaling Lymphocyte Activation Molecule) or CDw150, belongs to the immunoglobulin gene superfamily and is involved in T cell stimulation. SLAM is constitutively expressed on peripheral blood memory T cells, T cell clones, immature thymocytes, and a proportion of B cells, and is rapidly induced on naive T cells after activation. Activated B cells express the membrane bound form, the soluble form and the cytoplasmic isoforms of SLAM. It is suggested that signalling through homophilic SLAM-SLAM binding during B to B and B to T cell interactions enhances the expansion and differentiation of activated B cells. Reports suggest that the extracellular domain of CD150 is the receptor for the measles virus and acts as a co-activator on T cells and B cells.Specifications
| SLAM/CD150 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13291 | |
| SLAMF1 | |
| SLAM/CD150 Fusion Protein Ag1386 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| Western Blot | |
| 5B4G1 | |
| Unconjugated | |
| SLAMF1 | |
| CD150, CDw150, IPO 3, SLAM, SLAMF1 | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 6504 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction