missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAM/CD150 Antibody (SLAM.4), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62198UV
This item is not returnable.
View return policy
Description
SLAM/CD150 Monoclonal antibody specifically detects SLAM/CD150 in Human samples. It is validated for Immunoprecipitation, Immunocytochemistry
Specifications
| SLAM/CD150 | |
| Monoclonal | |
| DyLight 350 | |
| CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen | |
| Human CD150-transfected 300.19 cells | |
| 0.1 mL | |
| Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 6504 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunoprecipitation, Immunocytochemistry | |
| SLAM.4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction