missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLAM/CD150 Polyclonal antibody specifically detects SLAM/CD150 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | SLAM/CD150 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human SLAMF1. The immunogen is located within amino acids 30 - 80 of SLAMF1. Amino Acid Squence: LPLTYERINKSMNKS |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?