missing translation for 'onlineSavingsMsg'
Learn More

SLAM/CD150 Antibody (8), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18382891 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382891 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382891 Supplier Novus Biologicals Supplier No. NBP289678AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLAM/CD150
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SLAM/CD150 (NP_003028.1; Met 1-Pro 258).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6504
Test Specificity No cross-reactivity in ELISA with: Human CD48/ BCM1/ BLAST1 Human CD84/ SLAMF5 Human 2B4/ NKR2B4/ CD244
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.