missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAM/CD150 Antibody (8), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89678AF350
This item is not returnable.
View return policy
Description
SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SLAM/CD150 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SLAM/CD150 (NP_003028.1; Met 1-Pro 258). | |
| 0.1 mL | |
| Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
| 6504 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 8 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human CD48/ BCM1/ BLAST1 Human CD84/ SLAMF5 Human 2B4/ NKR2B4/ CD244 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction