missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAM/CD150 Antibody (542301), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1642CL1
This item is not returnable.
View return policy
Description
SLAM/CD150 Monoclonal antibody specifically detects SLAM/CD150 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,CyTOF
Specifications
| SLAM/CD150 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen | |
| Mouse myeloma cell line NS0-derived recombinant human SLAM/CD150, Ala21-Pro237, Accession # Q13291 | |
| 0.1 mL | |
| Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
| 6504 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Western Blot, CyTOF | |
| 542301 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction