missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLAIN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566605
This item is not returnable.
View return policy
Description
Immunogen sequence: SPARGIEYSRV SPQPMISRLQ QPRLSLQGHP TDLQTSNVKN EEKLRRSLPN LSRTSNTQVD SVKSSRSDSN FQVPN Highest antigen sequence identity to the following orthologs - mouse 97%, rat 99%.
SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
Specifications
| SLAIN2 | |
| Polyclonal | |
| Unconjugated | |
| SLAIN2 | |
| 5033405K12Rik; 8030444K12Rik; AI596370; FLJ21611; hypothetical protein LOC534203; KIAA1458; LOW QUALITY PROTEIN: SLAIN motif-containing protein 2; RGD1311593; SLAIN motif family member 2; SLAIN motif family, member 2; SLAIN motif-containing protein 2; SLAIN2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57606 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P270 | |
| SLAIN2 | |
| Recombinant Human SLAIN2. Recombinant protein control fragment (Product #RP-107260). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction