missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLAIN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572685
This item is not returnable.
View return policy
Description
SLAIN1 Polyclonal Antibody for IHC (P), ELISA
SLAIN1 (SLAIN Motif Family Member 1) is a Protein Coding gene. An important paralog of this gene is SLAIN2. Microtubule plus-end tracking protein that might be involved in the regulation of cytoplasmic microtubule dynamics, microtubule organization and microtubule elongation.
Specifications
| SLAIN1 | |
| Polyclonal | |
| Unconjugated | |
| SLAIN1 | |
| 9630044O09Rik; AA675320; AW742596; C13orf32; RGD1308626; SLAIN motif family member 1; SLAIN motif family, member 1; SLAIN motif-containing protein 1; SLAIN1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 122060 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8ND83 | |
| SLAIN1 | |
| SLAIN1 antibody for a 16 amino acid synthetic peptide near the center of human SLAIN1. The immunogen is within amino acids 370 - 420 of SLAIN1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction