missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLAIN1 Polyclonal antibody specifically detects SLAIN1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SLAIN1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | SLAIN motif family, member 1 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 518 and 568 of human SLAIN Motif Family, Member 1 using the numbering given in entry NP_001035243.1 (GeneID 122060). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?