missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLA2 Monoclonal Antibody (OTI2G3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526061
This item is not returnable.
View return policy
Description
SLA2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Adapter protein, which negatively regulates T-cell receptor (TCR) signaling. Inhibits T-cell antigen-receptor induced activation of nuclear factor of activated T-cells. May act by linking signaling proteins such as ZAP70 with CBL, leading to a CBL dependent degradation of signaling proteins.
Specifications
| SLA2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H6Q3 | |
| SLA2 | |
| Full length human recombinant protein of SLA2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2G3 | |
| Unconjugated | |
| SLA2 | |
| A930009E21Rik; AI430952; C20orf156; MARS; Modulator of antigen receptor signaling; modulator of antigen receptor signaling MARS; RGD1562071; Sla2; SLAP2; SLAP-2; Src like adaptor 2; src-like adapter protein 2; Src-like adapter protein-2; src-like-adapter 2; Src-like-adaptor 2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84174 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction