missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79830
This item is not returnable.
View return policy
Description
SLA2 Polyclonal specifically detects SLA2 in Human samples. It is validated for Western Blot.
Specifications
| SLA2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C20orf156, FLJ21992, Modulator of antigen receptor signaling, SLAP-2MARS, SLAP2MGC49845, Src-like adapter protein 2, Src-like adapter protein-2, src-like-adapter 2, Src-like-adaptor 2 | |
| Rabbit | |
| 28 kDa | |
| 100 μL | |
| Immunology, Signal Transduction | |
| 84174 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_115590 | |
| SLA2 | |
| Synthetic peptide directed towards the middle region of human SLA2The immunogen for this antibody is SLA2. Peptide sequence WLYEGLSREKAEELLLLPGNPGGAFLIRESQTRRGSYSLSVRLSRPASWD. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Dog: 86%; Rat: 86%; Horse: 86%; Guinea pig: 86%; Sheep: 79%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction