missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLA/LP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532121
This item is not returnable.
View return policy
Description
Recommended positive controls: A431. Predicted reactivity: Rat (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SLA/LP (SEPSECS) converts O-phosphoseryl-tRNA(Sec) to selenocysteinyl-tRNA(Sec) required for selenoprotein biosynthesis.
Specifications
| SLA/LP | |
| Polyclonal | |
| Unconjugated | |
| SEPSECS | |
| 9130208G10; AA986712; D5Ertd135e; hypothetical protein LOC393123; liver-pancreas antigen; LP; O-phosphoseryl-tRNA(Sec) selenium transferase; PCH2D; Sec synthase; SecS; Selenocysteine synthase; selenocysteinyl-tRNA(Sec) synthase; Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase; SEPSECS; sep-tRNA:Sec-tRNA synthase; SLA; SLA/LP; SLA/LP autoantigen; sla/lpl; SLA-p35; Soluble liver antigen; soluble liver antigen/liver pancreas antigen; soluble liver antigen/liver pancreas antigen, like; Soluble liver antigen/liver pancreas antigen-like; tRNA(Ser/Sec)-associated antigenic protein; TRNP48; UGA suppressor tRNA-associated protein; UGA suppressor tRNA-associated protein homolog; zgc:55980 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51091 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9HD40 | |
| SEPSECS | |
| Recombinant protein encompassing a sequence within the center region of human SLA/LP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction