missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLA/LP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577134
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
SLA/LP (SEPSECS) converts O-phosphoseryl-tRNA(Sec) to selenocysteinyl-tRNA(Sec) required for selenoprotein biosynthesis.
Specifications
| SLA/LP | |
| Polyclonal | |
| Unconjugated | |
| SEPSECS | |
| 9130208G10; AA986712; D5Ertd135e; hypothetical protein LOC393123; liver-pancreas antigen; LP; O-phosphoseryl-tRNA(Sec) selenium transferase; PCH2D; Sec synthase; SecS; Selenocysteine synthase; selenocysteinyl-tRNA(Sec) synthase; Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase; SEPSECS; sep-tRNA:Sec-tRNA synthase; SLA; SLA/LP; SLA/LP autoantigen; sla/lpl; SLA-p35; Soluble liver antigen; soluble liver antigen/liver pancreas antigen; soluble liver antigen/liver pancreas antigen, like; Soluble liver antigen/liver pancreas antigen-like; tRNA(Ser/Sec)-associated antigenic protein; TRNP48; UGA suppressor tRNA-associated protein; UGA suppressor tRNA-associated protein homolog; zgc:55980 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 211006, 51091, 679383 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P6M7, Q9HD40 | |
| SEPSECS | |
| Recombinant full length Human SLA/LP. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Del en indholdskorrektion