missing translation for 'onlineSavingsMsg'
Learn More

SL 0101-1, MedChemExpress

Product Code. 16428977
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16428977

Brand: Medchem Express HY15237

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SL 0101-1 (SL0101), a kaempferol glycoside, isolated from the tropical plant F. refracta, is a cell-permeable, selective, reversible, ATP-competitive p90 Ribosomal S6 Kinase (RSK) inhibitor, with an IC50 of 89 nM. SL 0101-1 (SL0101) is a selective RSK1/2 inhibitor, with a Ki of 1 μM.

SL 0101-1 (SL0101), a kaempferol glycoside, isolated from the tropical plant F. refracta, is a cell-permeable, selective, reversible, ATP-competitive p90 Ribosomal S6 Kinase (RSK) inhibitor, with an IC50 of 89 nM. SL 0101-1 (SL0101) is a selective RSK1/2 inhibitor, with a Ki of 1 μM.

Specifications

Chemical Name or Material SL 0101-1
CAS 77307-50-7
Color White
Purity Grade Notes Research
Molecular Formula C25H24O12
Quantity 5 mg
Synonym SL0101
SMILES OC1=CC(O)=C(C(C(O[C@@H]2O[C@@H](C)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H]2O)=C(C3=CC=C(O)C=C3)O4)=O)C4=C1
Molecular Weight (g/mol) 516.45
Formula Weight 516.45
Percent Purity 96.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.