missing translation for 'onlineSavingsMsg'
Learn More

Skp2 Inhibitor C1, MedChemExpress

Product Code. 16433147
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16433147

Brand: Medchem Express HY16661

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Skp2 Inhibitor C1(SKPin C1) is a specific small molecule inhibitor of Skp2-mediated p27 degradation, selectively inhibited Skp2-mediated p27 degradation by reducing p27 binding through key compound-receptor contacts.

Skp2 Inhibitor C1(SKPin C1) is a specific small molecule inhibitor of Skp2-mediated p27 degradation, selectively inhibited Skp2-mediated p27 degradation by reducing p27 binding through key compound-receptor contacts.

Specifications

Chemical Name or Material Skp2 Inhibitor C1
CAS 432001-69-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H13BrN2O4S2
Quantity 50 mg
Synonym SKPin C1
Solubility Information DMSO : 20.83 mg/mL (44.76 mM; Need ultrasonic)
SMILES S=C(S/1)N(CC2=CN=CC=C2)C(C1=C\C(C=C(Br)C=C3)=C3OCC(O)=O)=O
Molecular Weight (g/mol) 465.34
Formula Weight 465.34
Percent Purity 96.2%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.