missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Skp2 Inhibitor C1, MedChemExpress
Description
Skp2 Inhibitor C1(SKPin C1) is a specific small molecule inhibitor of Skp2-mediated p27 degradation, selectively inhibited Skp2-mediated p27 degradation by reducing p27 binding through key compound-receptor contacts.
Specifications
Specifications
| Chemical Name or Material | Skp2 Inhibitor C1 |
| CAS | 432001-69-9 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C18H13BrN2O4S2 |
| Quantity | 10 mg |
| Synonym | SKPin C1 |
| Solubility Information | DMSO : 20.83 mg/mL (44.76 mM; Need ultrasonic) |
| SMILES | S=C(S/1)N(CC2=CN=CC=C2)C(C1=C\C(C=C(Br)C=C3)=C3OCC(O)=O)=O |
| Molecular Weight (g/mol) | 465.34 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?