missing translation for 'onlineSavingsMsg'
Learn More

Skp2 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18403804 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403804 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403804 Supplier Novus Biologicals Supplier No. NBP130077V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Skp2 Polyclonal antibody specifically detects Skp2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Skp2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CDK2/cyclin A-associated protein p45, cyclin A/CDK2-associated protein p45, Cyclin-A/CDK2-associated protein p45, FBL1, F-box protein Skp2, F-box/LRR-repeat protein 1, FBXL1MGC1366, FLB1, p45skp2, S-phase kinase-associated protein 2, S-phase kinase-associated protein 2 (p45)
Host Species Rabbit
Immunogen Antigen maps to a region between residue 1 and 50 of human S-phase kinase-associated protein 2 using the numbering given in entry NP_005974.2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6502
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.