missing translation for 'onlineSavingsMsg'
Learn More

SKLB4771, MedChemExpress

Product Code. 16485448
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485448 10 mg 10mg
16461627 50 mg 50mg
16428167 5 mg 5mg
16426046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485448 Supplier Medchem Express Supplier No. HY12960

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SKLB4771 is a novel potent and selective Flt3 inhibitor with IC50 of 10 nM; against FLT3-ITD-expressing MV4-11 cells with IC50 of 6 nM.

SKLB4771 is a novel potent and selective Flt3 inhibitor with IC50 of 10 nM

Specifications

Chemical Name or Material SKLB4771
CAS 1370256-78-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H27N7O3S2
Quantity 10 mg
Synonym FLT3-​IN-​1
Solubility Information DMSO : 33.33 mg/mL (61.99 mM; Need ultrasonic)
SMILES O=C(NC1=NN=C(SC2=C3C=CC(OCCCN4CCOCC4)=CC3=NC=N2)S1)NC5=CC=C(C)C=C5
Molecular Weight (g/mol) 537.66
Formula Weight 537.66
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.