missing translation for 'onlineSavingsMsg'
Learn More

SKL2001, MedChemExpress

Product Code. 16494516
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494516 100 mg 100mg
16453258 10 mg 10mg
16410926 200 mg 200mg
16409806 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494516 Supplier Medchem Express Supplier No. HY101085
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SKL2001 is an agonist of the Wnt/β-catenin pathway, with anti-cancer activity. SKL2001 stabilizes intracellular β-catenin via disruption of the Axin/β-catenin interaction.

SKL2001 is an agonist of the Wnt/β-catenin pathway, with anti-cancer activity. SKL2001 stabilizes intracellular β-catenin via disruption of the Axin/β-catenin interaction.

Specifications

Chemical Name or Material SKL2001
CAS 909089-13-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H14N4O3
Quantity 100 mg
Solubility Information DMSO : ≥ 96.66 mg/mL (337.63 mM)
SMILES O=C(C1=NOC(C2=CC=CO2)=C1)NCCCN3C=CN=C3
Molecular Weight (g/mol) 286.29
Formula Weight 286.29
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.