Learn More
Invitrogen™ SKIV2L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596363
Description
Immunogen sequence: MADAFGDELF SVFEGDSTTA AGTKKDKEKD KGKWKGPPGS ADKAGKRFDG KLQSESTNNG KNKRDVDFEG TDEPIFGKKP RIEESITEDL SLADLMPRVK VQSVETVEGC THEVALPAEE DYLPLKPRVG KAAKEYPFIL DAFQREAIQC VDNNQSVLVS AHTSAGKTVC AEYAIALALR EKQRVIFTSP IKALSNQKYR EMYEEFQDVG LMTGDVTINP TASCLVMTTE ILRSMLYRGS EVMREVAWVI FDEIHYMRDS ERGVVWEETI ILLPDNVHYV FLSATIPNAR QFAEWICHLH KQPC; Positive Samples: HeLa, 22RV1, 293T, U-87MG, Mouse testis, Mouse spleen; Cellular Location: Nucleus, nucleolus, nucleoplasm.
SKIV2L2 may be involved in pre-mRNA splicing.
Specifications
| SKIV2L2 | |
| Polyclonal | |
| Unconjugated | |
| MTREX | |
| 2610528A15Rik; ATP-dependent helicase SKIV2L2; ATP-dependent RNA helicase DOB1; ATP-dependent RNA helicase SKIV2L2; Dob1; Exosome RNA helicase MTR4; exosome RNA helicase MTR4; superkiller viralicidic activity 2-like 2; fSAP118; functional spliceosome-associated protein 118; KIAA0052; mKIAA0052; Mtr4; Mtr4 exosome RNA helicase; MTREX; RGD1305984; Ski2 like RNA helicase 2; SKIV2L2; Superkiller viralicidic activity 2-like 2; superkiller viralicidic activity 2-like 2 (S. cerevisiae); superkiller viralicidic activity 2-like 2-like; TRAMP-like complex helicase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23517, 72198 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P42285, Q9CZU3 | |
| MTREX | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-304 of human SKIV2L2 (NP_0561753). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.