missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKIV2L2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93932-0.1ml
This item is not returnable.
View return policy
Description
SKIV2L2 Polyclonal antibody specifically detects SKIV2L2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SKIV2L2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ATP-dependent helicase SKIV2L2, Dob1, EC 3.6.1, EC 3.6.4.13, fSAP118, KIAA0052functional spliceosome-associated protein 118, MGC142069, Mtr4superkiller viralicidic activity 2-like 2, superkiller viralicidic activity 2-like 2 (S. cerevisiae) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human SKIV2L2 (NP_056175.3). MADAFGDELFSVFEGDSTTAAGTKKDKEKDKGKWKGPPGSADKAGKRFDGKLQSESTNNGKNKRDVDFEGTDEPIFGKKPRIEESITEDLSLADLMPRVKVQSVETVEGCTHEVALPAEEDYLPLKPRVGKAAKEYPFILDAFQREAIQCVDNNQSVLVSAHTSAGKTVCAEYAIALALREKQRVIFTSPIKALSNQKYREMYEEFQDVGLMTGDVTINPTASCLVMTTEILRSMLYRGSEVMREVAWVIFDEIHYMRDSERGVVWEETIILLPDNVHYVFLSATIPNARQFAEWICHLH | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23517 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction