missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SKIV2L Monoclonal Antibody (K94011_4C6), Invitrogen™

Mouse Monoclonal Antibody

£380.00 - £570.00

Specifications

Antigen SKIV2L
Clone K94011_4C6
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30212490
View Documents
Invitrogen™
MA556090
200 μL
£570.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30208876
View Documents
Invitrogen™
MA555710
50 μL
£380.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence of this protein is as follows: SEFPSRKDSK AHEQALAELT KRLGALEEPD MTGQLVDLPE YYSWGEELTE TQHMIQRRIM ESVNGLKSLS AGRVVVVKNQ EHHNALGVIL QVSSNSTSRV FTTLVLCDKP LSQDPQDRGP ATAEVPYPDD LVGFKLFLPE GPCDHTVVKL QPGDMAAITT KVLRVNGEKI LEDFSKRQQP KFKKDPPLAA VTTAVQELLR LAQAHPAGPP TLDPVNDLQL KDMSVVEGGL RARKLEELIQ GAQCVHSPRF PAQYLKLRER MQIQKEMERL

SKIV2L Monoclonal antibody specifically detects SKIV2L in Human samples. It is validated for Western Blot
Specifications

Specifications

SKIV2L
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
170A; 4930534J06Rik; AW214248; Ddx13; Helicase SKI2W; helicase-like protein; HLP; SKI; SKI2; SKI2 homolog, superkiller viralicidic activity 2-like; Ski2 like RNA helicase; Ski2w; SKIV2; SKIV2L; SKIV2L1; superkiller viralicidic activity 2-like; superkiller viralicidic activity 2-like (S. cerevisiae); THES2; W
Recombinant human SKIC2 (immunogen range is 771-1040).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
SKIV2L
K94011_4C6
Western Blot
Unconjugated
Mouse
Human
Q15477
6499
IgG2b
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.