missing translation for 'onlineSavingsMsg'
Learn More

SKIV2L Antibody, DyLight 550, Novus Biologicals™

Product Code. 18412288 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412288 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412288 Supplier Novus Biologicals Supplier No. NBP281903R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SKIV2L Polyclonal antibody specifically detects SKIV2L in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKIV2L
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias 170A, DDX13helicase SKI2W, EC 3.6.1, Helicase-like protein, HLPEC 3.6.4.-, SKI2, SKI2Wsuperkiller viralicidic activity 2 (S. cerevisiae homolog)-like, SKIV2, superkiller viralicidic activity 2-like (S. cerevisiae), W
Host Species Rabbit
Immunogen SKIV2L antibody was raised against a 16 amino acid peptide near the amino terminus of human SKIV2L. The immunogen is located within amino acids 60 - 110 of SKIV2L. Amino Acid Squence: EHSARKWQRKTDPWSL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Unfolded Protein Response, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6499
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.