Learn More
Invitrogen™ SKIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596816
Description
Immunogen sequence: HLPPHISNNY QRLEHFDRIL EMQNCEGRDI PNILDHDLII WFGDMNFRIE DFGLHFVRES IKNRCYGGLW EKDQLSIAKK HDPLLREFQE GRLLFPPTYK FDRNSNDYDT SEKKRKPAWT DRILWRLKRQ PCAGPDTPIP PASHFSLSLR GYSSHMTYGI SDHKPVSGTF DLELKPLVSA PLIVLMPEDL WTVENDMMVS YSSTSDFPSS PWDWIGLYKV GLRDVNDYVS YAWVGDSKVS CSDNLNQVYI DISNIPTTED EFLLCYYSNS LRSVVGISRP FQIPPGSLRE DPLGEAQPQI; Positive Samples: Mouse brain, Rat brain; Cellular Location: Endoplasmic reticulum.
This gene encodes a protein with 5-phosphatase activity toward polyphosphate inositol. The protein localizes to the cytosol in regions lacking actin stress fibers. It is thought that this protein may negatively regulate the actin cytoskeleton. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| SKIP | |
| Polyclonal | |
| Unconjugated | |
| INPP5K | |
| C62; Inositol polyphosphate 5-phosphatase K; inositol polyphosphate-5-phosphatase K; Inpp5k; Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase; Phosphatidylinositol-4,5-bisphosphate 5-phosphatase; PI-5-phosphatase related; PPS; putative phosphoinositide 5-phosphatase type II; putative phosphoinositide 5-phosphatase type II; C62; putative PI-5-phosphatase; skeletal muscle and kidney enriched inositol phosphatase; skeletal muscle and kidney enriched inositol polyphosphate phosphatase; skeletal muscle and kidney-enriched inositol phosphatase; SKIP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19062, 287533, 51763 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8C5L6, Q9BT40 | |
| INPP5K | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 149-448 of human INPP5K (NP_0576162). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.