Learn More
Tork Skincare Dispenser S4
Product Code. 30243859 Shop All Tork Products
Change view
Click to view available options
Color:
Black
Silver
White
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243859 Silver Each
30243836 Black Each
30243818 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30243859 Supplier Tork Supplier No. 460010
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Tork Skincare Dispenser fits any location and can be used with a wide range of Tork soaps, sanitisers and lotions in sealed bottles.
  • Easy-to-use design promotes hand hygiene for all (certified by the Swedish Rheumatism Association, tested according to The Design for All Test, published in the Swedish Design Research Journal 2011).
  • Help secure good hand hygiene - the factory-sealed bottle with a new pump for every refill helps reduce the risk of contamination and protect the formulations all the way to the hands.
  • Save staff time with effortless refilling in less than 10 seconds (based on internal panel test).
  • High capacity reduces waste and helps reduce consumption by up to 50% (based on Essity test using Tork Foam Soap vs Tork Liquid Soap in Elevation dispenser).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hand Cleanser Dispenser
Color Silver
Alternate Part Number 7322540782004
Holds Hand Sanitizer, Hand Soap, Hand Lotion
Material Plastic
Mounting Wall Mount
No. of Compartments 1
Product Style Hinged Lid
Model Image