missing translation for 'onlineSavingsMsg'
Learn More

Skimmin, MedChemExpress

Product Code. 16499268
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499268 10 mg 10mg
16471698 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499268 Supplier Medchem Express Supplier No. HYN2263
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Skimmin (Umbelliferone glucoside) is a coumarin found in Hydrangea paniculata, inhibits immune complex deposition, with anti-inflammatory activity.

Skimmin (Umbelliferone glucoside) is a coumarin found in Hydrangea paniculata, inhibits immune complex deposition, with anti-inflammatory activity.

Specifications

Chemical Name or Material Skimmin
CAS 93-39-0
Color White
Purity Grade Notes Research
Molecular Formula C15H16O8
Quantity 10 mg
Synonym Umbelliferone glucoside
Solubility Information DMSO : 100 mg/mL (308.38 mM; Need ultrasonic)
SMILES O=C1C=CC2=CC=C(O[C@H]3[C@@H]([C@H]([C@@H]([C@@H](CO)O3)O)O)O)C=C2O1
Molecular Weight (g/mol) 324.28
Formula Weight 324.28
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.