missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKI II, Tocris Bioscience™
Description
Selective non-lipid inhibitor of sphingosine kinase (IC50 = 0.5 μM); does not act at ATP-binding site. Displays no inhibition of ERK2, PI 3-kinase, or PKCα at concentrations up to 60 μM. Reduces levels of sphingosine-1-phosphate in MDA-MB-231 breast cancer cells. Induces apoptosis and inhibits proliferation in several other tumor cell lines in vitro (IC50 = 0.9 - 4.6 μM).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 302.78 |
| Percent Purity | >98% |
| Chemical Name or Material | SKI II |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?