missing translation for 'onlineSavingsMsg'
Learn More

SKI II, Tocris Bioscience™

Product Code. 18024882
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18024882 10 mg 10mg
18045192 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18024882 Supplier Tocris Bioscience Supplier No. 2097/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective non-lipid inhibitor of sphingosine kinase

Selective non-lipid inhibitor of sphingosine kinase (IC50 = 0.5 μM); does not act at ATP-binding site. Displays no inhibition of ERK2, PI 3-kinase, or PKCα at concentrations up to 60 μM. Reduces levels of sphingosine-1-phosphate in MDA-MB-231 breast cancer cells. Induces apoptosis and inhibits proliferation in several other tumor cell lines in vitro (IC50 = 0.9 - 4.6 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312636-16-1
Molecular Formula C15H11ClN2OS
Molecular Weight (g/mol) 302.776
InChI Key ZFGXZJKLOFCECI-UHFFFAOYSA-N
Synonym ski ii, 4-4-4-chlorophenyl-2-thiazolyl amino phenol, skiii, sphk-i2, 4-4-4-chlorophenyl-1,3-thiazol-2-yl amino phenol, sphingosine kinase inhibitor 2, 4-4-4-chlorophenyl thiazol-2-ylamino phenol, phenol, 4-4-4-chlorophenyl-2-thiazolyl amino, 3vzd
PubChem CID 753704
IUPAC Name 4-[[4-(4-chlorophenyl)-1,3-thiazol-2-yl]amino]phenol
SMILES C1=CC(=CC=C1C2=CSC(=N2)NC3=CC=C(C=C3)O)Cl

Specifications

Quantity 10 mg
Formula Weight 302.78
Percent Purity >98%
Chemical Name or Material SKI II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.