missing translation for 'onlineSavingsMsg'
Learn More

SKI II, MedChemExpress

Product Code. 16493386
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493386 25 mg 25mg
16491106 200 mg 200mg
16485878 10 mg 10mg
16482027 50 mg 50mg
16426346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493386 Supplier Medchem Express Supplier No. HY13822
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SKI-II is an oral active and synthetic inhibitor of sphingosine kinase (SK) activity, with IC50 values of 78 μM and 45 μM for SK1 and for SK2, respectively. SKI II causes an irreversible inhibition of SK1 by inducing its lysosomal and/or proteasomal degradation.

SKI-II is an oral active and synthetic inhibitor of sphingosine kinase (SK) activity, with IC50 values of 78 μM and 45 μM for SK1 and for SK2, respectively. SKI II causes an irreversible inhibition of SK1 by inducing its lysosomal and/or proteasomal degradation.

Specifications

Chemical Name or Material SKI II
CAS 312636-16-1
Color White
Purity Grade Notes Research
Molecular Formula C15H11ClN2OS
Quantity 25 mg
Solubility Information DMSO : ≥ 100 mg/mL (330.27 mM)
SMILES OC1=CC=C(NC2=NC(C3=CC=C(Cl)C=C3)=CS2)C=C1
Molecular Weight (g/mol) 302.78
Formula Weight 302.78
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.