missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SKI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29937
This item is not returnable.
View return policy
Description
SKI Polyclonal antibody specifically detects SKI in Human samples. It is validated for Immunoprecipitation
Specifications
| SKI | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6497 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| C-ski, Proto-oncogene c-Ski, ski oncogene, ski oncoprotein, SKV, v-ski avian sarcoma viral oncogene homolog, v-ski sarcoma viral oncogene homolog (avian) | |
| between 450 and 500 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction