missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKI Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £375.00
Specifications
| Antigen | SKI |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628022
|
Novus Biologicals
NBP2-94105-0.02ml |
0.02 mL |
£159.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659551
|
Novus Biologicals
NBP2-94105-0.1ml |
0.1 mL |
£375.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SKI Polyclonal antibody specifically detects SKI in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SKI | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| C-ski, Proto-oncogene c-Ski, ski oncogene, ski oncoprotein, SKV, v-ski avian sarcoma viral oncogene homolog, v-ski sarcoma viral oncogene homolog (avian) | |
| A synthetic peptide corresponding to a sequence within amino acids 650-728 of human SKI (NP_003027.1). CDKGCEAGRLRAKYSAQIEDLQVKLQHAEADREQLRADLLREREAREHLEKVVKELQEQLWPRARPEAAGSEGAAELEP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 6497 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title