missing translation for 'onlineSavingsMsg'
Learn More

SKF89976A hydrochloride, MedChemExpress

Product Code. 16429516
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16429516

Brand: Medchem Express HY100228A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SKF89976A hydrochloride is a selective GABA transporter (GAT-1) inhibitor with IC50s of 0.28 μM, 137.34 μM and 202.8 μM for GAT-1, GAT-2 and GAT-3 in CHO cells, respectively.

SKF89976A hydrochloride is a selective GABA transporter (GAT-1) inhibitor with IC50s of 0.28 μM, 137.34 μM and 202.8 μM for GAT-1, GAT-2 and GAT-3 in CHO cells, respectively.

Specifications

Chemical Name or Material SKF89976A hydrochloride
CAS 85375-15-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H26ClNO2
Quantity 50 mg
Synonym d,l-SKF89976A hydrochloride
Solubility Information DMSO : 100 mg/mL (268.89 mM; Need ultrasonic) ∣H2O : 20 mg/mL (53.78 mM; Need ultrasonic)
SMILES O=C(C1CN(CC/C=C(C2=CC=CC=C2)/C3=CC=CC=C3)CCC1)O.[H]Cl
Molecular Weight (g/mol) 371.9
Formula Weight 371.9
Percent Purity 98.72%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.