missing translation for 'onlineSavingsMsg'
Learn More

SKF89976A hydrochloride, MedChemExpress

Product Code. 16495597
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495597 5 mg 5mg
16472858 10 mg 10mg
16429516 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495597 Supplier Medchem Express Supplier No. HY100228A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SKF89976A hydrochloride is a selective GABA transporter (GAT-1) inhibitor with IC50s of 0.28 μM, 137.34 μM and 202.8 μM for GAT-1, GAT-2 and GAT-3 in CHO cells, respectively.

SKF89976A hydrochloride is a selective GABA transporter (GAT-1) inhibitor with IC50s of 0.28 μM, 137.34 μM and 202.8 μM for GAT-1, GAT-2 and GAT-3 in CHO cells, respectively.

Specifications

Chemical Name or Material SKF89976A hydrochloride
CAS 85375-15-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H26ClNO2
Quantity 5 mg
Synonym d,l-SKF89976A hydrochloride
Solubility Information DMSO : 100 mg/mL (268.89 mM; Need ultrasonic) ∣H2O : 20 mg/mL (53.78 mM; Need ultrasonic)
SMILES O=C(C1CN(CC/C=C(C2=CC=CC=C2)/C3=CC=CC=C3)CCC1)O.[H]Cl
Molecular Weight (g/mol) 371.9
Formula Weight 371.9
Percent Purity 98.72%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.