missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SKF 97541

Product Code. 18056032
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18056032

Brand: Tocris Bioscience™ 0379/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Highly potent GABAB agonist; also GABAA-ρ antagonist

SKF 97541 is a potent GABAB agonist (EC50 = 50 nM). SKF 97541 depresses striatal synaptic potentials and hyperpolarizes nigral neurons (EC50 values = 92 nM and 150 nM, respectively). SKF 97541 shows antidepressant effects in animal models and is also a GABAA-ρ antagonist.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127729-35-5
Molecular Formula C4H12NO2P

Specifications

Quantity 50 mg
Formula Weight 137.12
Purity >98%
Molecular Weight (g/mol) 137.119
Chemical Name or Material SKF 97541
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.