missing translation for 'onlineSavingsMsg'
Learn More

SKF-96365 hydrochloride, MedChemExpress

Product Code. 16485467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485467 5 mg 5mg
16462728 10 mg 10mg
16460896 200 mg 200mg
16459426 50 mg 50mg
16414196 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16485467 Supplier Medchem Express Supplier No. HY1000015mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SKF-96365 hydrochloride is a potent TRP channel blocker and a store-operated Ca2+ entry (SOCE) inhibitor. SKF-96365 hydrochloride significantly inhibits hERG, hKCNQ1/hKCNE1, hKir2.1 and hKv4.3 current, and significantly prolongs the QTc interval in isolated guinea pig hearts. SKF-96365 hydrochloride exhibits potent anti-neoplastic activity by inducing cell-cycle arrest and apoptosis in colorectal cancer cells.

SKF-96365 hydrochloride is a potent TRP channel blocker and a store-operated Ca2+ entry (SOCE) inhibitor. SKF-96365 hydrochloride significantly inhibits hERG, hKCNQ1/hKCNE1, hKir2.1 and hKv4.3 current, and significantly prolongs the QTc interval in isolated guinea pig hearts. SKF-96365 hydrochloride exhibits potent anti-neoplastic activity by inducing cell-cycle arrest and apoptosis in colorectal cancer cells.

Specifications

Chemical Name or Material SKF-96365 hydrochloride
CAS 130495-35-1
Color White
Purity Grade Notes Research
Molecular Formula C22H27ClN2O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (248.19 mM; Need ultrasonic) ∣H2O : 100 mg/mL (248.19 mM; Need ultrasonic)
SMILES COC1=CC=C(CCCOC(C2=CC=C(OC)C=C2)CN3C=CN=C3)C=C1.[H]Cl
Molecular Weight (g/mol) 402.91
Formula Weight 402.91
Percent Purity 99.51%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.