missing translation for 'onlineSavingsMsg'
Learn More

SKF-82958 hydrobromide, MedChemExpress

Product Code. 16493968
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493968 10 mg 10mg
16440327 50 mg 50mg
16436687 5 mg 5mg
16402936 25 mg 25mg
16494976 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493968 Supplier Medchem Express Supplier No. HY10435A
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SKF-82958 ((±)-SKF 82958) hydrobromide is a dopamine D1 receptor full agonist (K0.5=4 nM), displays selective for D1 over D2 receptors (K0.5=73 nM). SKF-82958 hydrobromide induces dopamine D1 receptor-dependent adenylate cyclase activity in rat striatal membranes (EC50=491 nM).

SKF-82958 ((±)-SKF 82958) hydrobromide is a dopamine D1 receptor full agonist (K0.5=4 nM), displays selective for D1 over D2 receptors (K0.5=73 nM). SKF-82958 hydrobromide induces dopamine D1 receptor-dependent adenylate cyclase activity in rat striatal membranes (EC50=491 nM).

Specifications

Chemical Name or Material SKF-82958 hydrobromide
CAS 74115-01-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H21BrClNO2
Quantity 10 mg
Synonym (±)-SKF-82958 hydrobromideChloro-APB hydrobromide
Solubility Information DMSO : ≥ 100 mg/mL (243.47 mM)
SMILES OC1=C(O)C=C2C(C3=CC=CC=C3)CN(CC=C)CCC2=C1Cl.[H]Br
Molecular Weight (g/mol) 410.73
Formula Weight 410.73
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.