missing translation for 'onlineSavingsMsg'
Learn More

SKF 81297C, TRC

Product Code. 30426143
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426143 100 mg 100mg
30408994 10 mg 10mg
30426142 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426143 Supplier Toronto Research Chemicals Supplier No. TRCS550020100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67287-39-2
Molecular Formula C16H17BrClNO2
Molecular Weight (g/mol) 370.67
Synonym 6-Chloro-2,3,4,5-tetrahydro-1-phenyl-1H-3-benzazepine-7,8-diol Hydrobromide, 6-Chloro-1-phenyl-2,3,4,5-tetrahydro-1H-benzo[d]azepine-7,8-diol Hydrobromide, SKF 81297 Hydrobromide
IUPAC Name 6-chloro-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol;hydrobromide
SMILES OC1=CC2=C(C(Cl)=C1O)CCNCC2C3=CC=CC=C3.Br

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H16ClNO2.BrH/c17-15-11-6-7-18-9-13(10-4-2-1-3-5-10)12(11)8-14(19)16(15)20;/h1-5,8,13,18-20H,6-7,9H2;1H
Formula Weight 369.0131
Chemical Name or Material SKF 81297C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.