missing translation for 'onlineSavingsMsg'
Learn More

SKF 38393 hydrochloride, MedChemExpress

Product Code. 16411046
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16411046

Brand: Medchem Express HY12520A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SKF 38393 hydrochloride is a selective agonist of the dopamine D1 receptor (D1DR) with an IC50 of 110 nM.

SKF 38393 hydrochloride is a selective agonist of the dopamine D1 receptor (D1DR) with an IC50 of 110 nM.

Specifications

Chemical Name or Material SKF 38393 hydrochloride
CAS 62717-42-4
Color Brown
Purity Grade Notes Research
Molecular Formula C16H18ClNO2
Quantity 200 mg
Synonym (±)-SKF-38393 hydrochloride SKF-38393A
Solubility Information DMSO : ≥ 34 mg/mL (116.53 mM) ∣H2O : 10 mg/mL (34.27 mM; Need ultrasonic)
SMILES OC1=C(O)C=C2C(C3=CC=CC=C3)CNCCC2=C1.[H]Cl
Molecular Weight (g/mol) 291.77
Formula Weight 291.77
Percent Purity 98.22%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.