missing translation for 'onlineSavingsMsg'
Learn More

Skepinone-L, MedChemExpress

Product Code. 16492537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492537 50 mg 50mg
16449027 5 mg 5mg
16446438 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492537 Supplier Medchem Express Supplier No. HY15300
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Skepinone-L (CBS3830) is a selective p38 mitogen-activated protein kinase inhibitor.

Skepinone-L (CBS3830) is a selective p38 mitogen-activated protein kinase inhibitor.

Specifications

Chemical Name or Material Skepinone-L
CAS 1221485-83-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H21F2NO4
Quantity 50 mg
Synonym CBS3830
Solubility Information DMSO : ≥ 50 mg/mL (117.53 mM)
SMILES O=C1C2=CC(OC[C@@H](CO)O)=CC=C2CCC3=C1C=CC(NC4=CC=C(C=C4F)F)=C3
Molecular Weight (g/mol) 425.42
Formula Weight 425.42
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.