Learn More
Honeywell™ SKELETON NIT 3 Gloves
Product Code. 15816785
Click to view available options
Size:
8
9
10
Unit Size:
1 pair
Product Code. 15816785
Brand: Honeywell™ 233227510
Only null left
Add to basket
Add to basket
Honeywell™ SKELETON NIT 3 Gloves feature a shock absorber pad on top of the hand to protect metacarpian and fingers.
  • Knitted glove in Spectra™
  • The knitting technique provides the glove with excellent fit and high dexterity
  • The Spectra™ yarn offers very good resistance to cut
  • Black nitrile foam coating on palm
  • Nitrile coating provides good protection against oil and grease
  • The foam coating makes the glove more flexible than standard nitrile
  • Rubber pad on top of the hand and fingers
  • Helps to protect metacarpian and fingers from shocks
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards
Description 13 gauge
Color Blue, White
Material Nitrile
Size 10
Length (Metric) 25 cm